Learn More
Chemglass Life Sciences Coupling Assembly Complete, Split Coupling, 30mm, Universal, Stainless Bushing and Upper Adapter For Use with CG-2033-B-25, CG-2033-B-38, & CG-2033-B39
SDP
Catalog No. 501531422
Encompass_Preferred
Coupling Assembly Complete, Split Coupling, 30mm, Universal, Stainless Bushing and Upper Adapter For Use with CG-2033-B-25. Split coupling design allows for easier assembly and disassembly between the motor and 30mm glass stirrer shaft. Coupling with universal joint compensates for misalignment of drive motor and glass stir shaft. Nylon bushing compresses around shaft using an allen screw. Stainless Steel collar/bushing and upper coupling attach directly to the motor shaft. For use with 100L process reactor systems.
Catalog No. 50-153-1422 Supplier Chemglass Life Sciences Supplier No. CG2046X130
Add to Cart
Add to Cart
Coupling Assembly Complete, Split Coupling, 30mm, Universal, Stainless Bushing and Upper Adapter For Use with CG-2033-B-25. Split coupling design allows for easier assembly and disassembly between the motor and 30mm glass stirrer shaft. Coupling with universal joint compensates for misalignment of drive motor and glass stir shaft. Nylon bushing compresses around shaft using an allen screw. Stainless Steel collar/bushing and upper coupling attach directly to the motor shaft. For use with 100L process reactor systems.