Learn More
Chemglass Life Sciences 25.4mm Stirrer Shaft Coupling, For Use With CG-2033-X-11 Motor, Gold Motor Adapter
SDP
Catalog No. 50872497
Encompass_Preferred
25.4mm Stirrer Shaft Coupling, For Use With CG-2033-X-11 Motor, Gold Motor Adapter. Incorporates a universal stirrer shaft coupling (CG-2046) to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Stainless steel stud and upper coupling attach directly to the motor shaft. For use with 30L thru 100L process reactor systems.
Catalog No. 50-872-497 Supplier Chemglass Life Sciences Supplier No. CG204692
25.4mm Stirrer Shaft Coupling, For Use With CG-2033-X-11 Motor, Gold Motor Adapter. Incorporates a universal stirrer shaft coupling (CG-2046) to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Stainless steel stud and upper coupling attach directly to the motor shaft. For use with 30L thru 100L process reactor systems.