Learn More
Chemglass Life Sciences 30mm Stirrer Shaft Coupling, For Use With CG-2025-20 Motor, Aqua Marine Motor Adapter
SDP
Catalog No. 501531420
Encompass_Preferred
30mm Stirrer Shaft Coupling, For Use With CG-2025-20 Motor, Aqua Marine Motor Adapter. Incorporates a universal stirrer shaft coupling (CG-2046) to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Stainless Steel stud and upper coupling attach directly to the motor shaft. For use with 30L thru 100L process reactor systems.
Catalog No. 50-153-1420 Supplier Chemglass Life Sciences Supplier No. CG204696
30mm Stirrer Shaft Coupling, For Use With CG-2025-20 Motor, Aqua Marine Motor Adapter. Incorporates a universal stirrer shaft coupling (CG-2046) to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Stainless Steel stud and upper coupling attach directly to the motor shaft. For use with 30L thru 100L process reactor systems.