Learn More
Wilmad Labglass COUPLING BEAD FIT TFE/VTN 1IN
Catalog No. 50800390
Encompass
Coupling, Beaded Fitting, PTFE/Viton Fits Fitting ID: 1 in. Provides insert and greaseless connection For equipment having Beaded fittings. Design eliminates pressure points which cause glassware to break.
Catalog No. 50-800-390 Supplier Wilmad Labglass Supplier No. LG4661100
Coupling, Beaded Fitting, PTFE/Viton Fits Fitting ID: 1 in. Provides insert and greaseless connection For equipment having Beaded fittings. Design eliminates pressure points which cause glassware to break.