Learn More
Wilmad Labglass COUPLING BEAD FIT TFE/VTN 2IN
Catalog No. 50800451
Encompass
Coupling, Beaded Fitting, PTFE/Viton Fits Fitting ID: 2 in. Provides insert and greaseless connection For equipment having Beaded fittings. Design eliminates pressure points which cause glassware to break.
Catalog No. 50-800-451 Supplier Wilmad Labglass Supplier No. LG4661104
Coupling, Beaded Fitting, PTFE/Viton Fits Fitting ID: 2 in. Provides insert and greaseless connection For equipment having Beaded fittings. Design eliminates pressure points which cause glassware to break.