Learn More
TA Instruments Coupling Dual Light Guide Assembly, Provides Secure and Efficient Connection Between Light Source and PCA or DSC Cell, Optimizes Light Transmission
Catalog No. 5000580612 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Coupling Dual Light Guide Assembly ensures secure, stable, and efficient transmission of light from the UV source to the PCA or DSC cell. This assembly provides optimized optical alignment, minimizing energy loss and ensuring consistent intensity at the target. Designed for durability and ease of use, it allows precise integration into photocalorimetry and curing setups. Its robust construction and compatibility make it a reliable accessory for extending the performance and usability of dual light guides in laboratory environments.
Catalog No. 50-005-80612 Supplier TA Instruments Supplier No. 935036.901
Waters TA Instruments Coupling Dual Light Guide Assembly ensures secure, stable, and efficient transmission of light from the UV source to the PCA or DSC cell. This assembly provides optimized optical alignment, minimizing energy loss and ensuring consistent intensity at the target. Designed for durability and ease of use, it allows precise integration into photocalorimetry and curing setups. Its robust construction and compatibility make it a reliable accessory for extending the performance and usability of dual light guides in laboratory environments.