Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) HFCD17835 Quick-disconnect fittings, Polysulfone valved hose barbs, 1/2""
Catalog No. 50627721
Encompass
High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately). Plug insert into coupling body to connect¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 SS latch springs and EPDM seals.
Catalog No. 50-627-721 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3130711
High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately). Plug insert into coupling body to connect¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 SS latch springs and EPDM seals.