Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) LCD16004 Metal Quick-Disconnect Panel-Mount Body; Valved, 1/4"" ID, 1/4"" flow
Catalog No. NC1441908
Encompass
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.
Catalog No. NC1441908 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3140372
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.