Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMC1702 Quick-Disconnect Hose Barb Fitting Body; Straight-through, 1/8"" Tube ID
Catalog No. 50628150
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-150 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636111
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.