Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPC32003 All-Plastic Quick Disconnect Sealing Cap, PC, 5/pack
Catalog No. 50627889
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-889 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131164
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).