Learn More
Wilmad Labglass COUPLING FLEXIBLE 10MM 5/16IN
Supplier:  Wilmad Labglass LG9598110
Encompass
Coupling, Flexible, w/ PTFE Sleeve. Stirring Shaft Size: 10mm; Motor Shaft Size: 3/8 in. 4 in. length allows For compensation of misalignment. Rubber sleeve body provides quiet, vibration free running.
Catalog No. 50-800-332
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.