Learn More
Grainger Coupling Forged Steel 1/2 in FNPT
Catalog No. 502750610
Encompass_Preferred
The Anvil Forged Steel Coupling is a high-quality industrial coupling made in the USA from forged carbon steel. It has a maximum pressure rating of 300 psi and temperature rating of 300°F. The coupling has NPT thread connections and is available in sizes ranging from 1/8" to 2". It can be used to join pipes, valves, and fittings in various industrial, commercial, and residential applications. The heavy-duty forged construction provides durability and strength while the short length allows for easy installation in tight spaces. This coupling is an ideal choice for creating strong, leak-free connections in plumbing, HVAC, hydraulic, and other fluid transfer systems.
Catalog No. 50-275-0610 Supplier Grainger Supplier No. 14J739
The Anvil Forged Steel Coupling is a high-quality industrial coupling made in the USA from forged carbon steel. It has a maximum pressure rating of 300 psi and temperature rating of 300°F. The coupling has NPT thread connections and is available in sizes ranging from 1/8" to 2". It can be used to join pipes, valves, and fittings in various industrial, commercial, and residential applications. The heavy-duty forged construction provides durability and strength while the short length allows for easy installation in tight spaces. This coupling is an ideal choice for creating strong, leak-free connections in plumbing, HVAC, hydraulic, and other fluid transfer systems.