Learn More
Grainger Coupling Forged Steel 3/4 in FNPT
Catalog No. 502749524
Encompass_Preferred
The Anvil Forged Steel Coupling is a high-quality industrial coupling made in the USA from forged carbon steel. It has a maximum pressure rating of 300 psi and temperature rating of 300°F. The coupling has NPT thread connections and is available in sizes ranging from 1/8" to 2". It can be used to join pipes, valves, and fittings in various industrial, commercial, and residential applications. The heavy-duty forged construction provides durability and strength.
Catalog No. 50-274-9524 Supplier Grainger Supplier No. 14J740
The Anvil Forged Steel Coupling is a high-quality industrial coupling made in the USA from forged carbon steel. It has a maximum pressure rating of 300 psi and temperature rating of 300°F. The coupling has NPT thread connections and is available in sizes ranging from 1/8" to 2". It can be used to join pipes, valves, and fittings in various industrial, commercial, and residential applications. The heavy-duty forged construction provides durability and strength.