Learn More
World Precision Instrument PolyFil Multi-Barrel Micropipette Coupling Kit
SDP
Catalog No. 50821829
Encompass
PolyFil™ allows easy and secure coupling of a multi-barrel micropipette to a pres­sure source. Cou­pling is achieved by bonding tem­per­a­ture-re­sis­tant and flexible MicroFil to the capillary tube with hot melt ad­he­sive. The luer end of each MicroFil is connected to PVC tubing (200 PSI rated). Kits also include a five-port manifold that allows use of a sin­gle PV830 Series PicoPump to drive up to six micropipette barrels independently by switching on only the barrels to be injected. All connections are locking luers - pressure safe and convenient.
Catalog No. 50-821-829 Supplier World Precision Instrument Supplier No. 5440
PolyFil™ allows easy and secure coupling of a multi-barrel micropipette to a pres­sure source. Cou­pling is achieved by bonding tem­per­a­ture-re­sis­tant and flexible MicroFil to the capillary tube with hot melt ad­he­sive. The luer end of each MicroFil is connected to PVC tubing (200 PSI rated). Kits also include a five-port manifold that allows use of a sin­gle PV830 Series PicoPump to drive up to six micropipette barrels independently by switching on only the barrels to be injected. All connections are locking luers - pressure safe and convenient.