Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPX22839M All-Plastic Quick Disconnect Barbed Inserts; Polysulfone (PS), 1/2"" Tube ID, 5/pack
Catalog No. 50627920
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-920 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131173
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).