Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD2203 Quick-Disconnect Hose Barb Insert; Valved, 3/16"" Tube ID, Acetal
Catalog No. 50628251
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-251 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636043
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.