Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PLCD22005 Quick-Disconnect Hose Barb Insert; Valved, 5/16"" Tube ID, 1/4"" Flow Size
Catalog No. 50628105
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-105 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636083
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.