Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPC30039M All-Plastic Quick Disconnect Sealing Plug, Polysulfone (PS), 5/pack
Catalog No. 50627887
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
This item is not returnable. View return policy
Catalog No. 50-627-887 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131178
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).