Learn More
Ace Glass, Inc. COUPLING NYLON 15MM COUPLING NYLON 15MM
SDP
Catalog No. 50972278
Encompass_Preferred
15mm nylon coupling for coupling threaded columns together, or coupling threaded columns to end fittings with a single o-ring seal for leak-tight engagement, with hand pressure and no significant size reduction in I.D. Supplied with two FETFE o-rings.
Catalog No. 50-972-278 Supplier Ace Glass, Inc. Supplier No. 584010
15mm nylon coupling for coupling threaded columns together, or coupling threaded columns to end fittings with a single o-ring seal for leak-tight engagement, with hand pressure and no significant size reduction in I.D. Supplied with two FETFE o-rings.