Learn More
Ace Glass, Inc. COUPLING NYLON 50MM COUPLING NYLON 50MM
SDP
Catalog No. 50972279
Encompass_Preferred
50mm nylon coupling for coupling threaded columns together, or coupling threaded columns to end fittings with a single o-ring seal for leak-tight engagement, with hand pressure and no significant size reduction in I.D. Supplied with two FETFE o-rings.
Catalog No. 50-972-279 Supplier Ace Glass, Inc. Supplier No. 584020
50mm nylon coupling for coupling threaded columns together, or coupling threaded columns to end fittings with a single o-ring seal for leak-tight engagement, with hand pressure and no significant size reduction in I.D. Supplied with two FETFE o-rings.