Learn More
Ace Glass, Inc. COUPLING PTFE 50MM COUPLING PTFE 50MM
SDP
Catalog No. 50972282
Encompass_Preferred
50mm PTFE coupling for coupling threaded columns together, or coupling threaded columns to end fittings with a single o-ring seal for leak-tight engagement, with hand pressure and no significant size reduction in I.D. Supplied with two FETFE o-rings.
Catalog No. 50-972-282 Supplier Ace Glass, Inc. Supplier No. 584051
50mm PTFE coupling for coupling threaded columns together, or coupling threaded columns to end fittings with a single o-ring seal for leak-tight engagement, with hand pressure and no significant size reduction in I.D. Supplied with two FETFE o-rings.