Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPCK17004T39 Quick-Disconnect Body with Lock, PS, 1/4"" ID; 5/Pk
Catalog No. 50627900
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-900 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131126
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).