Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPCK17006T03 Quick-Disconnect Body with Lock; PC, 3/8"" ID; 5/Pk
Catalog No. 50627902
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-902 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131117
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).