Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPCK17006T39 Quick-Disconnect Body with Lock, PS, 3/8"" ID; 5/Pk
Catalog No. 50627904
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-904 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131127
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).