Learn More
COLDER PRODUCTS COMPANY CPC HFC35 Polysulfone Hose Barb Coupling Bodies In-line coupling Free floating EPDM seal options Vacuum to 125 psi 8 6 bar Valve Spring 316 Stainless Steel
Catalog No. 503131398
Encompass_Preferred
CPC HFC35 Polysulfone Hose Barb Coupling Bodies In-line coupling Free floating EPDM seal options Vacuum to 125 psi 8 6 bar Valve Spring 316 Stainless Steel
Catalog No. 50-313-1398 Supplier COLDER PRODUCTS COMPANY Supplier No. HFCD17835
CPC HFC35 Polysulfone Hose Barb Coupling Bodies In-line coupling Free floating EPDM seal options Vacuum to 125 psi 8 6 bar Valve Spring 316 Stainless Steel