Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMC1304 Quick-Disconnect Body; Straight-through, 1/4"" tube OD, 1/8"" flow, acetal
Catalog No. 50628142
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-142 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636108
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.