Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PLCD13006 Compression Fitting Body; Valved, 3/8"" Tube OD, 1/4"" Flow Size, Acetal
Catalog No. 50628070
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-070 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0635991
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.