Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD1703 Hose Barb Fitting Body; Valved, 3/16"" Tube ID, 1/8"" Flow Size, Acetal
Catalog No. 50628227
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-227 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636022
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.