Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPXK17803 Quick-Disconnect Body with Lock; PC, 1/2"" ID; 5/Pk
Catalog No. 50627925
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-925 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131118
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).