Learn More
Grainger Grote Hi Count LED Courtesy Light, Clear, 12V, 1 ea
Catalog No. 502771797
Encompass_Preferred
The Hi Count™ LED courtesy and utility lamp is designed for interior wall or ceiling applications where space is limited. This lamp utilizes LED technology to provide efficient illumination while maintaining a low profile.
  • Provides maximum light output in tight spaces
  • Features low power consumption and heat production for efficient operation
  • Equipped with a hooded guard to reduce glare
  • Constructed with an acrylic lens and steel housing
  • Designed for 12V DC systems
  • Surface mount installation via screw connection
  • Includes blunt cut electrical connection
Catalog No. 50-277-1797 Supplier Grainger Supplier No. 13X012
The Hi Count™ LED courtesy and utility lamp is designed for interior wall or ceiling applications where space is limited. This lamp utilizes LED technology to provide efficient illumination while maintaining a low profile.
  • Provides maximum light output in tight spaces
  • Features low power consumption and heat production for efficient operation
  • Equipped with a hooded guard to reduce glare
  • Constructed with an acrylic lens and steel housing
  • Designed for 12V DC systems
  • Surface mount installation via screw connection
  • Includes blunt cut electrical connection