Learn More
Creative Biomart Recombinant CoV-2 N (1-419) Protein, His-tagged
Supplier:  Creative Biomart COV2366
Encompass_Preferred
The E. coli derived recombinant protein contains the Coronavirus 2019 CoV-2 Nucleocapsid Phosphoprotein, Wuhan-Hu-1 strain, amino acids (1-419) having a Mw of 46.4 kDa and fused to 6 His tag at C-t
Catalog No. 50-208-4744