Learn More
Creative Biomart Recombinant CoV-2 N (201-419) Protein, His-tagged
Supplier:  Creative Biomart COV2369
Encompass_Preferred
The E. coli derived recombinant protein contains the Coronavirus 2019 C-Terminal domain nuclepocapsid (201-419 a,a.) fused to 6 His tag at C-terminal and having a molecular weight of 23.9 kDa.
Catalog No. 50-208-4747