Learn More
Creative Biomart Recombinant CoV-2-S1 Protein, His-tagged
Supplier:  Creative Biomart COV2386
Encompass_Preferred
The HEK293 derived recombinant protein contains the Coronavirus 2019 Spike Glycoprotein S1 Receptor Binding Domain [RBD], Wuhan-Hu-1 strain, amino acids 319-541 fused to His tag at C-terminal.
Catalog No. 50-208-4764