Learn More
Creative Biomart Recombinant CoV-2 S1 (319-537) Protein, His-tagged
Supplier:  Creative Biomart COV2375
Encompass_Preferred
The HEK293 derived recombinant protein contains the Coronavirus 2019 Spike Glycoprotein S1 Receptor Binding Domain [RBD], Wuhan-Hu-1 strain, amino acids 319-537, having a Mw of 26.5 kDa. Cov-2 RBD is
Catalog No. 50-208-4753