Learn More
Creative Biomart Recombinant CoV-2 S1 Protein, His-tagged
Supplier:  Creative Biomart COV2376
Encompass_Preferred
The Sf9 derived recombinant protein contains the Coronavirus 2019 CoV-2 Spike Glycoprotein S1, Wuhan-Hu-1 strain, amino acids 1-674 fused to His tag at C-terminal.
Catalog No. 50-208-4754