Learn More
Creative Biomart Recombinant CoV-2 S2 Protein, His-tagged
Supplier:  Creative Biomart COV2378
Encompass_Preferred
The Sf9 derived recombinant protein contains the Coronavirus 2019 CoV-2 Spike Glycoprotein S2, Wuhan-Hu-1 strain, 685-1211 amino acids, having a Mw of 60.1 kDa and fused to 6 His tag at C-terminal.
Catalog No. 50-208-4756