Learn More
Creative Biomart Recombinant CoV-2-Spike Protein, Fc-tagged
Supplier:  Creative Biomart COV2390
Encompass_Preferred
The HEK293 derived recombinant protein contains the Coronavirus 2019-Spike N-Terminal Domain, Wuhan-Hu-1 strain, amino acids 1-260 fused to Fc tag at C-terminal.
Catalog No. 50-208-4768