Learn More
Creative Biomart Porcine Circovirus Type 2 Gene Engineering Subunit Vaccine Antigen
Supplier:  Creative Biomart COV2384
Encompass_Preferred
Porcine circovirus type 2 (PCV2) can cause Postweaning multisystemic wasting syndrome(PMWS), porcine dermatitis and nephropathy syndrome (PDNS), porcine respiratory disease complex(PRDC), piglet reproductive dysfunction (PRF), piglet congenital tremors (CT) and a series of disease, and easy to mix with other pathogen infection or lead to secondary infection, increased the complexity of clinical. The Porcine Circovirus Type 2 Gene Engineering Subunit Vaccine has multiple immune functions, such as humoral immunity and cellular immunity; And is safer and more efficient because it contains no nucleic acid, with pure antigen and high content; It is a pure virus-like particle circosubunit vaccine; Immunity once, protection all the way.
Catalog No. 50-208-4762