Learn More
Sino Biological CoV Spike MAb 20ul (Sino Biological)
Supplier:  Sino Biological 68086MM2320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant CoV spike S1+S2 ECD protein. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1847