Learn More
Bollé Safety Covaclave Safety Goggles
Catalog No. 18072135
Change view
Click to view available options
Lens Coating:
Scratch-resistant
Catalog No. Lens Coating
18-072-135 Scratch-resistant
1 options
Catalog No. 18-072-135 Supplier Bollé Safety Supplier No. COVACLAVE
Add to Cart
Add to Cart
Covaclave safety goggles developed to guarantee high protection and comfort in risky environments, like pharmaceutics, micro-electronics or surgery departments
  • Overflow chute for liquids
  • Indirect and interchangeable ventilation
  • High-comfort autoclavable goggles
  • Fit over prescription glasses
  • Polycarbonate lens without coating
  • Isoprene strap
  • Autoclave for sterilization
Specifications
Lens Coating Scratch-resistant
Shape Rectangular
Certifications/Compliance ANSI Z87+
Lens Tint Clear
Product Type Safety Goggles
Frame Color Clear
Headband Material Isoprene Strap
Weight (English) 3.4 oz.
Disposable Reusable
Lens Material Polycarbonate
Frame Material TPR
UV Protection UVA and UVB
Frame Type Vented