Learn More
Bollé Safety Covarsi Safety Goggles
Catalog No. 18072134
Change view
Click to view available options
Frame Type:
Sealed
Vented
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Type
18-072-134 Vented
18-072-133 Sealed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-072-134 Supplier Bollé Safety Supplier No. COVARSI
Add to Cart
Edge
Add to Cart
Covarsi safety goggles maximum safety for all including wearers of prescription glasses, polycarbonate, OTG, features high wrap around
  • Economical and effective
  • Comfortable, flexible frame
  • Adjustable strap with pivoting buckle
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87+
Shape Rectangular
Lens Tint Clear
Product Type Safety Goggles
Frame Color Clear
Weight (English) 3.42 oz.
Disposable Reusable
Lens Material Polycarbonate
UV Protection UVA and UVB
Frame Type Vented