Learn More
Grainger Cove Cut Profile Router Bit 7/8
Catalog No. 502766277
Encompass_Preferred
The Westward Cove Cut Profile Router Bit is a 1/4" shank router bit with a 1-1/2" cutting diameter. It features a carbide tipped 1/4" shank and a 1/2" cutting height. This cove cut router bit creates a concave profile and is designed for use on wood, plywood, and laminates. It has a maximum RPM of 24,000. The overall length of the bit is 2-1/4". This router bit allows you to add decorative edges and cut cove profiles.
Catalog No. 50-276-6277 Supplier Grainger Supplier No. 16Y606
The Westward Cove Cut Profile Router Bit is a 1/4" shank router bit with a 1-1/2" cutting diameter. It features a carbide tipped 1/4" shank and a 1/2" cutting height. This cove cut router bit creates a concave profile and is designed for use on wood, plywood, and laminates. It has a maximum RPM of 24,000. The overall length of the bit is 2-1/4". This router bit allows you to add decorative edges and cut cove profiles.