Learn More
Chemtronics™ Coventry™ Ultrajet™ Dusters
Catalog No. 19003233
Change view
Click to view available options
Volume (English):
8 oz.
10 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-003-233 10 oz.
19-035-235 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-003-233 Supplier Chemtronics™ Supplier No. ES1020
Add to Cart
Edge
Add to Cart
For applications that require safe, ultra-pure cleaning
Compact and portable source of canned air. Microscopically clean, oil- and moisture-free.
  • Removes particles instantly from the most inaccessible areas
  • Economize solvent usage by removing layers of dry particulate and allowing the solvent to work immediately of encrusted soils
  • Accelerate surface drying when using slow evaporating cleaners to remove damaging oil and residue
  • Ultrajet All-Way offers the advantage of spraying in any direction with no danger of a liquid spray that might freeze or damage delicate components
Specifications
Product Type Gas Duster
Flammable No
For Use With (Application) Removing dust build up from circuit boards, cleaning insulating debris from pin connectors, removal of entrapped solvent from under surface mount devices
Volume (English) 10 oz.
Volume (Metric) 283 g