Learn More
Hach Company SC200 UV Protection Screen
Catalog No. 501997279 Shop All Hach Company Products
Encompass
This assembly covers the face of the SC200 Universal Controller, protecting it from the sun. The transparent door can be opened for operating the controller. It is UV absorbent and protects the display and membrane keypad from deterioration due to UV exposure. It will become yellowish over time. This door is available as a replacement part (9221700). Controller included in picture for display purposes only.
Catalog No. 50-199-7279 Supplier Hach Company Supplier No. 8809200
This assembly covers the face of the SC200 Universal Controller, protecting it from the sun. The transparent door can be opened for operating the controller. It is UV absorbent and protects the display and membrane keypad from deterioration due to UV exposure. It will become yellowish over time. This door is available as a replacement part (9221700). Controller included in picture for display purposes only.