Learn More
Electron Microscopy Sciences Easy-Load Cover for Biopsy Cassettes, White
SDP
Catalog No. 5030330
Encompass_Preferred
One-piece disposable plastic, it can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss.
Color: White
Catalog No. 50-303-30 Supplier Electron Microscopy Sciences Supplier No. 70063W
One-piece disposable plastic, it can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss.
Color: White