Learn More
Med Vet International Dressing Retention Tape With Liner 3M Medipore Dress-It White 3-7/8 X 7-7/8 inch Soft Cloth Nonsterile, 2955, 100/Case
Catalog No. 502831306
Encompass_Preferred
3M Medipore Dress-It pre-cut dressing cover secures dressings where skin may stretch It comes pre-cut to save time and effort The material is gentle and breathable to enhance comfort and maintain the skin It's stretchy to help conform easily around body contours The 3-7/8-inch x 7-7/8-inch cloth tape provides no self-adhesion or curling, making it easy to handle It is latex-free to prevent allergic reactions Each box includes 25 non-sterile white pads
Catalog No. 50-283-1306 Supplier Med Vet International Supplier No. 1054279CS
3M Medipore Dress-It pre-cut dressing cover secures dressings where skin may stretch It comes pre-cut to save time and effort The material is gentle and breathable to enhance comfort and maintain the skin It's stretchy to help conform easily around body contours The 3-7/8-inch x 7-7/8-inch cloth tape provides no self-adhesion or curling, making it easy to handle It is latex-free to prevent allergic reactions Each box includes 25 non-sterile white pads