Learn More
Med Vet International Dressing Retention Tape With Liner 3M Medipore Dress-It White 5-7/8 X 5-7/8 inch Soft Cloth Nonsterile, 2956, 25/Box
Catalog No. 502831373
Encompass_Preferred
The non-sterile 3M Medipore Dress-It pre-cut dressing cover secures dressings where skin may stretch It comes pre-cut to save time and effort The material is gentle and breathable to enhance comfort and maintain the skin It's stretchy to help conform easily around body contours The 5-7/8-inch x 5-7/8-inch cloth tape provides no self-adhesion or curling, making it easy to handle The white tape is latex-free to prevent allergic reactions Each pad includes three sheets
Catalog No. 50-283-1373 Supplier Med Vet International Supplier No. 684046BX
The non-sterile 3M Medipore Dress-It pre-cut dressing cover secures dressings where skin may stretch It comes pre-cut to save time and effort The material is gentle and breathable to enhance comfort and maintain the skin It's stretchy to help conform easily around body contours The 5-7/8-inch x 5-7/8-inch cloth tape provides no self-adhesion or curling, making it easy to handle The white tape is latex-free to prevent allergic reactions Each pad includes three sheets