Learn More
Med Vet International Dressing Retention Tape With Liner 3M Medipore Dress-It White 7-7/8 X 11 inch Soft Cloth Nonsterile, 2958, 100/Case
Catalog No. 502831372
Encompass_Preferred
The 3M Medipore Dress-It pre-cut dressing cover secures dressings where skin may stretch It comes pre-cut to save time and effort The material is gentle and breathable to enhance comfort and maintain the skin It's stretchy to help conform easily around body contours The 7-7/8-inch x 11-inch cloth tape provides no self-adhesion or curling, making it easy to handle The white tape is latex-free to prevent allergic reactions There are 25 non-sterile pads in a box
Catalog No. 50-283-1372 Supplier Med Vet International Supplier No. 683385CS
The 3M Medipore Dress-It pre-cut dressing cover secures dressings where skin may stretch It comes pre-cut to save time and effort The material is gentle and breathable to enhance comfort and maintain the skin It's stretchy to help conform easily around body contours The 7-7/8-inch x 11-inch cloth tape provides no self-adhesion or curling, making it easy to handle The white tape is latex-free to prevent allergic reactions There are 25 non-sterile pads in a box