Learn More
Med Vet International Dressing Retention Tape With Liner 3M Medipore Dress-It White 5-7/8 X 11 inch Soft Cloth Nonsterile, 2957, 100/Case
Catalog No. 502832054
Encompass_Preferred
3M Medipore Dress-It pre-cut dressing cover secures dressings where skin may stretch It comes pre-cut to save time and effort The material is gentle and breathable to enhance comfort and maintain the skin It's stretchy to help conform easily around body contours The 5-7/8-inch x 11-inch cloth tape provides no self-adhesion or curling, making it easy to handle
Catalog No. 50-283-2054 Supplier Med Vet International Supplier No. 209701CS
3M Medipore Dress-It pre-cut dressing cover secures dressings where skin may stretch It comes pre-cut to save time and effort The material is gentle and breathable to enhance comfort and maintain the skin It's stretchy to help conform easily around body contours The 5-7/8-inch x 11-inch cloth tape provides no self-adhesion or curling, making it easy to handle