Learn More
Kuhner Shaker Inc COVER FOR 24 DEEPWELL PLATE P
Catalog No. NC1212197
Encompass
Duetz Sandwich cover for 24 DWP with spongy silicone, fixed with pins, 1.2 mm holes, low evaporation
Catalog No. NC1212197 Supplier Kuhner Shaker Inc Supplier No. SMCR1224C
Duetz Sandwich cover for 24 DWP with spongy silicone, fixed with pins, 1.2 mm holes, low evaporation